missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzyl alcohol, TRC

Product Code. 30386469
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity unitSize
30386469 10 g 10g
1 options
This item is not returnable. View return policy

Product Code. 30386469

Brand: Toronto Research Chemicals TRCD43969010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3,4-Dichlorophenyl)methanol
CAS 1805-32-9
Molecular Formula C7 H6 Cl2 O
Quantity 10 g
Synonym 3,4-Dichlorobenzenemethanol, (3,4-Dichlorophenyl)methanol, 3,4-Dichlorobenzyl alcohol, NSC 26136, NSC 407829, 2,4-Dichlorobenzyl alcohol Imp. C (EP)
InChI Formula InChI=1S/C7H6Cl2O/c8-6-2-1-5(4-10)3-7(6)9/h1-3,10H,4H2
SMILES OCc1ccc(Cl)c(Cl)c1
IUPAC Name (3,4-dichlorophenyl)methanol
Molecular Weight (g/mol) 177.03
Formula Weight 175.9796
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.