missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorophenylacetonitrile, TRC

Product Code. 30369088
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30369088 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30369088

Brand: Toronto Research Chemicals TRCD4354355G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,4-Dichlorophenylacetonitrile
CAS 3218-49-3
Molecular Formula C8H5Cl2N
Quantity 5 g
Synonym (3,4-Dichlorophenyl)acetonitrile, 2-(3,4-Dichlorophenyl)acetonitrile, 3,4-Dichlorobenzeneacetonitrile, 3,4-Dichlorobenzyl cyanide, NSC 111725
InChI Formula InChI=1S/C8H5Cl2N/c9-7-2-1-6(3-4-11)5-8(7)10/h1-2,5H,3H2
SMILES N#CCC1=CC(Cl)=C(Cl)C=C1
IUPAC Name 2-(3,4-dichlorophenyl)acetonitrile
Molecular Weight (g/mol) 186.04
Formula Weight 184.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.