missing translation for 'onlineSavingsMsg'
Learn More

3-[4-(Trifluoromethyl)phenyl]propionic acid, 97%, Thermo Scientific™

Catalog Number p-7035802
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
53473-36-2
C10H9F3O2
218.175
MFCD00674032
OEIUMLSCWINLBB-UHFFFAOYSA-N
4-trifluoromethyl hydrocinnamic acid, 3-4-trifluoromethyl phenyl propanoic acid, 3-4-trifluoromethyl phenyl propionic acid, benzenepropanoic acid, 4-trifluoromethyl, 3-4-trifluoromethylphenyl propionic acid, 4-trifluoromethyl phenylpropionic acid, p-trifluoromethyl benzylacetic acid, 3-4-trifluoromethyl-phenyl-propionic acid
3588736
3-[4-(trifluoromethyl)phenyl]propanoic acid
C1=CC(=CC=C1CCC(=O)O)C(F)(F)F
This item is not returnable. View return policy
53473-36-2
C10H9F3O2
218.175
MFCD00674032
OEIUMLSCWINLBB-UHFFFAOYSA-N
4-trifluoromethyl hydrocinnamic acid, 3-4-trifluoromethyl phenyl propanoic acid, 3-4-trifluoromethyl phenyl propionic acid, benzenepropanoic acid, 4-trifluoromethyl, 3-4-trifluoromethylphenyl propionic acid, 4-trifluoromethyl phenylpropionic acid, p-trifluoromethyl benzylacetic acid, 3-4-trifluoromethyl-phenyl-propionic acid
3588736
3-[4-(trifluoromethyl)phenyl]propanoic acid
C1=CC(=CC=C1CCC(=O)O)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 99°C
Quantity 1 g
Formula Weight 218.17
Percent Purity 97%
Chemical Name or Material 3-[4-(Trifluoromethyl)phenyl]propionic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'