missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Di(tert-butyl)-4-hydroxybenzonitrile, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 1 g |
IUPAC Name | 3,5-ditert-butyl-4-hydroxybenzonitrile |
Formula Weight | 231.34 |
Percent Purity | 97% |
Chemical Name or Material | 3, 5-Di(tert-butyl)-4-hydroxybenzonitrile |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'