Learn More
3,5-dichlorobenzene-1-carbohydrazide, 97%, Thermo Scientific™
Catalog Number p-4391292
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
62899-78-9
C7H6Cl2N2O
205.04
MFCD00833411
DNISXKBKTSHNKL-UHFFFAOYSA-N
3,5-dichlorobenzhydrazide, 3,5-dichlorobenzenecarbohydrazide, acmc-1b4zh, 3,5-bis chloranyl benzohydrazide, 3,5-dichlorobenzoic acid hydrazide, benzoic acid,3,5-dichloro-,hydrazide, benzoic acid,3,5-dichloro-, hydrazide
2735966
3,5-dichlorobenzohydrazide
NNC(=O)C1=CC(Cl)=CC(Cl)=C1
62899-78-9
C7H6Cl2N2O
205.04
MFCD00833411
DNISXKBKTSHNKL-UHFFFAOYSA-N
3,5-dichlorobenzhydrazide, 3,5-dichlorobenzenecarbohydrazide, acmc-1b4zh, 3,5-bis chloranyl benzohydrazide, 3,5-dichlorobenzoic acid hydrazide, benzoic acid,3,5-dichloro-,hydrazide, benzoic acid,3,5-dichloro-, hydrazide
2735966
3,5-dichlorobenzohydrazide
NNC(=O)C1=CC(Cl)=CC(Cl)=C1
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 205.04
Percent Purity 97%
Chemical Name or Material 3, 5-dichlorobenzene-1-carbohydrazide