missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoro-4-(trifluoromethyl)benzoic acid, 98%, Thermo Scientific™

Catalog Number p-7053599
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
261945-09-9
C8H3F5O2
226.10
MFCD01631619
MKESWMDKZFRERU-UHFFFAOYSA-N
23443746
3,5-difluoro-4-(trifluoromethyl)benzoic acid
OC(=O)C1=CC(F)=C(C(F)=C1)C(F)(F)F
This item is not returnable. View return policy
261945-09-9
C8H3F5O2
226.10
MFCD01631619
MKESWMDKZFRERU-UHFFFAOYSA-N
23443746
3,5-difluoro-4-(trifluoromethyl)benzoic acid
OC(=O)C1=CC(F)=C(C(F)=C1)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Synonym 3,5-difluoro-4-trifluoromethyl benzoic acid, 3, 5-difluoro-4-trifluoromethyl benzoic acid, pubchem12004, benzoic acid,3,5-difluoro-4-trifluoromethyl, benzoic acid, 3,5-difluoro-4-trifluoromethyl, 3,5-bis fluoranyl-4-trifluoromethyl benzoic acid
Formula Weight 226.1
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3,5-Difluoro-4-(trifluoromethyl)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'