missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxybenzylamine, 98%

Product Code. p-7040671
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
15416978 5 g 5g
15426978 25 g 25g
2 options
This item is not returnable. View return policy

Product Code. 15416978

Brand: Thermo Scientific Alfa Aesar H60503.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 34967-24-3 | C9H13NO2 | 167.208 g/mol

3,5-Dimethoxybenzylamine was used in the preparation of trisammonium tris(hexafluoro phosphate) salt.  It was also used in the synthesis of well defined, homogeneous [n]rotaxanes (n up to 11) by a template-directed thermodynamic clipping approach.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethoxybenzylamine was used in the preparation of trisammonium tris(hexafluoro phosphate) salt.  It was also used in the synthesis of well defined, homogeneous [n]rotaxanes (n up to 11) by a template-directed thermodynamic clipping approach.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents. Store under room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34967-24-3
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.208
MDL Number MFCD00052813
InChI Key YGZJTYCCONJJGZ-UHFFFAOYSA-N
Synonym 3,5-dimethoxybenzylamine, 3,5-dimethoxybenzyl amine, 3,5-dimethoxyphenyl methanamine, 1-3,5-dimethoxyphenyl methanamine, 3,5-dimethoxy benzyl amine, 3,5-dimethoxyphenyl methylamine, benzenemethanamine, 3,5-dimethoxy, pubchem7393, 3,5-dimethoxy benzylamine, 3,5-dimethoxy-benzylamine
PubChem CID 420973
IUPAC Name (3,5-dimethoxyphenyl)methanamine
SMILES COC1=CC(=CC(=C1)CN)OC

Specifications

Melting Point 35°C to 37°C
Density 1.106
Boiling Point 94°C to 96°C
Flash Point 113°C (235°F)
Refractive Index 1.542
Quantity 5 g
UN Number UN3259
Solubility Information Slightly soluble in water.
Formula Weight 167.21
Percent Purity 98%
Chemical Name or Material 3,5-Dimethoxybenzylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.