missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-4-nitropyridine-2-methanol, 98%, Thermo Scientific™

Product Code. 11328766
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11328766 25 g 25g
11318766 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11328766

Brand: Thermo Scientific Alfa Aesar L19452.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149082-03-1
Molecular Formula C8H10N2O3
Molecular Weight (g/mol) 182.179
MDL Number MFCD03426035
InChI Key KBCDOXSSYLFMHH-UHFFFAOYSA-N
Synonym 3,5-dimethyl-4-nitropyridin-2-yl methanol, 3,5-dimethyl-2-hydroxymethyl-4-nitropyridine, 3,5-dimethyl-4-nitropyridine-2-methanol, 3,5-dimethyl-4-nitro-2-pyridinemethanol, 2-hydroxymethyl-3,5-dimethyl-4-nitropyridine, 3,5-dimethyl-4-nitro-2-pyridyl-1-methanol, 2-pyridinemethanol, 3,5-dimethyl-4-nitro, 3,5-dimethyl-4-nitro-2-pyridyl methan-1-ol, 4-nitro-3,5-dimethyl-2-hydroxymethyl pyridine
PubChem CID 4049415
IUPAC Name (3,5-dimethyl-4-nitropyridin-2-yl)methanol
SMILES CC1=CN=C(C(=C1[N+](=O)[O-])C)CO

Specifications

Melting Point 65°C to 68°C
Quantity 25 g
UN Number UN2811
Formula Weight 182.18
Percent Purity 98%
Chemical Name or Material 3,5-Dimethyl-4-nitropyridine-2-methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.