missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylisoxazole-4-boronic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-7029433
61.00 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
16114-47-9
C5H8BNO3
140.933
MFCD02677945
DIIFZCPZIRQDIJ-UHFFFAOYSA-N
3,5-dimethylisoxazole-4-boronic acid, 3,5-dimethylisoxazol-4-yl boronic acid, 3,5-dimethyl-4-isoxazolylboronic acid, 3,5-dimethylisoxazol-4-ylboronic acid, 3,5-dimethyl-1,2-oxazol-4-yl boronic acid, 3,5-dimethylisoxazole-4-boronicacid, 3,5-dimethyl-4-isoxazoleboronic acid, 3,5-dimethylisoxazol-4-yl-4-boronic acid, 3,5-dimethyl-isoxazole-4-boronic acid, dimethyl-1,2-oxazol-4-yl boronic acid
2734346
(3,5-dimethyl-1,2-oxazol-4-yl)boronic acid
B(C1=C(ON=C1C)C)(O)O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
16114-47-9
C5H8BNO3
140.933
MFCD02677945
DIIFZCPZIRQDIJ-UHFFFAOYSA-N
3,5-dimethylisoxazole-4-boronic acid, 3,5-dimethylisoxazol-4-yl boronic acid, 3,5-dimethyl-4-isoxazolylboronic acid, 3,5-dimethylisoxazol-4-ylboronic acid, 3,5-dimethyl-1,2-oxazol-4-yl boronic acid, 3,5-dimethylisoxazole-4-boronicacid, 3,5-dimethyl-4-isoxazoleboronic acid, 3,5-dimethylisoxazol-4-yl-4-boronic acid, 3,5-dimethyl-isoxazole-4-boronic acid, dimethyl-1,2-oxazol-4-yl boronic acid
2734346
(3,5-dimethyl-1,2-oxazol-4-yl)boronic acid
B(C1=C(ON=C1C)C)(O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 106°C to 108°C
Quantity 1 g
Formula Weight 140.93
Percent Purity 97%
Chemical Name or Material 3,5-Dimethylisoxazole-4-boronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'