Learn More
3,5-Dinitrobenzyl alcohol, 98%
Product Code. 15455417
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 15455417
Brand: Thermo Scientific Alfa Aesar H32669.06
Request Bulk or Custom Format
CAS: 71022-43-0 | C7H6N2O5 | 198.13 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 71022-43-0
Molecular Formula C7H6N2O5
Molecular Weight (g/mol) 198.13
MDL Number MFCD00007235
InChI Key GPHYIQCSMDYRGJ-UHFFFAOYSA-N
Synonym 3,5-dinitrobenzyl alcohol, 3,5-dinitrophenyl methanol, benzenemethanol, 3,5-dinitro, 3,5-dinitrobenzenemethanol, unii-111z6y5ynw, 3,5-dinitrobenzylalcohol, pubchem20861, acmc-1bdpg, ksc494c8n, 3,5-dinitrophenyl methanol #
PubChem CID 116978
IUPAC Name (3,5-dinitrophenyl)methanol
SMILES OCC1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O
Specifications
Melting Point 88°C to 91°C
Quantity 5 g
Beilstein 2054388
Formula Weight 198.13
Percent Purity 98%
Chemical Name or Material 3,5-Dinitrobenzyl alcohol