Learn More
3,6,9-Triethyl-3,6,9-trimethyl-1,4,7-triperoxonane, 41% solution in aromatic free mi, Thermo Scientific Chemicals
Product Code. p-203748 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
Product Code. 10534521
Brand: Thermo Scientific Acros 349940050
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Radical initiators
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3,6,9-Triethyl-3,6,9-trimethyl-1,4,7-triperoxonane
Color Undesignated
Name Note 41% Solution in Aromatic Free Min. Spirit
CAS 24748-23-0, 78-93-3
CAS Min % 40.0, 58.0
CAS Max % 42.0, 60.0
Melting Point <10.0°C
Molecular Formula C12H24O6
MDL Number MFCD02685709
Density 0.8750g/mL
Synonym unii-j3mv6azn38, 3,6,9-triethyl-3,6,9-trimethyl-1,2,4,5,7,8-hexoxonane, 1,2,4,5,7,8-hexoxonane, 3,6,9-triethyl-3,6,9-trimethyl, j3mv6azn38, 3,6,9-triethyl-3,6,9-trimethyl-1,4,7-triperoxynonane, unii-j3mv6azn38 component kvwlloiegklbpa-aqhstmncsa-n, unii-j3mv6azn38 component kvwlloiegklbpa-zsbigdgjsa-n, 3,6,9-triethyl-3,6,9-trimethyl-1,2,4,5,7,8-hexaoxacyclononane, 1,2,4,5,7,8-hexaoxacyclononane, 3,6,9-trimethyl, 3,6,9-triethyl, 3,6,9-triethyl-3,6,9-trimethyl-1,4,7-triperoxynonane sol.in arom.free min.spirit
Solubility Information Solubility in water: immiscible. Other solubilities: miscible with most organic solvents
InChI Key KVWLLOIEGKLBPA-UHFFFAOYSA-N
SMILES CCC1(OOC(OOC(OO1)(C)CC)(C)CC)C
IUPAC Name 3,6,9-triethyl-3,6,9-trimethyl-1,2,4,5,7,8-hexaoxonane
Molecular Weight (g/mol) 264.318
PubChem CID 2734078
Formula Weight 264.32
Percent Purity 7.3 to 7.6% (total active oxygen)
Packaging Plastic bottle
Specific Gravity 0.875
Viscosity 5 mPa.s (20°C)
Quantity 5 g
Flash Point 74°C
Physical Form Solution
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3,6,9-Triethyl-3,6,9-trimethyl-1,4,7-triperoxynonane, 41% solution in aromatic free mineral spirit
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Organic peroxide Type C & D
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H242-Heating may cause a fire.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P331-Do NOT induce vomiting.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P370+P380+P375-In case of fire: Evacuate area. Fight fire remotely due to the risk of explosion.
  • P403+P235-Store in a well-ventilated place. Keep cool.