missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dichloro-2-(trifluoromethyl)pyridine, 97%, Thermo Scientific™

Catalog Number p-4812895
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
89719-91-5
C6H2Cl2F3N
215.98
MFCD00525616
WDFIJOQQKFWJEA-UHFFFAOYSA-N
3,6-dichloro-2-trifluoromethyl pyridine, pyridine, 3,6-dichloro-2-trifluoromethyl, 2,5-dichloro-6-trifluoromethyl pyridine, acmc-20a3gu
20213813
3,6-dichloro-2-(trifluoromethyl)pyridine
FC(F)(F)C1=C(Cl)C=CC(Cl)=N1
This item is not returnable. View return policy
89719-91-5
C6H2Cl2F3N
215.98
MFCD00525616
WDFIJOQQKFWJEA-UHFFFAOYSA-N
3,6-dichloro-2-trifluoromethyl pyridine, pyridine, 3,6-dichloro-2-trifluoromethyl, 2,5-dichloro-6-trifluoromethyl pyridine, acmc-20a3gu
20213813
3,6-dichloro-2-(trifluoromethyl)pyridine
FC(F)(F)C1=C(Cl)C=CC(Cl)=N1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Boiling Point 88°C (16.0 mmHg)
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 215.99
Percent Purity 97%
Chemical Name or Material 3, 6-Dichloro-2-(trifluoromethyl)pyridine
compliance-icons
Product Identifier
  • 3,6-Dichloro-2-(trifluoromethyl)pyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'