missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dichloro-2-(trifluoromethyl)pyridine, 97%, ACROS Organics™

Brand:  Acros Organics 451380010

Additional Details : CAS : 89719-91-5 Weight : 0.01000kg


Quantity 1g

 View more versions of this product

Product Code. 15279756

  • 98.10€ / 1g


Estimated Shipment
31-07-2020
Add to basket


Specifications

Chemical Identifiers

89719-91-5
215.99
3,6-dichloro-2-trifluoromethyl pyridine, pyridine, 3,6-dichloro-2-trifluoromethyl, 2,5-dichloro-6-trifluoromethyl pyridine, acmc-20a3gu
3,6-dichloro-2-(trifluoromethyl)pyridine
C6H2Cl2F3N
WDFIJOQQKFWJEA-UHFFFAOYSA-N
20213813
C1=CC(=NC(=C1Cl)C(F)(F)F)Cl

Specifications

3, 6-Dichloro-2-(trifluoromethyl)pyridine
88°C (16.0mmHg)
89719-91-5
C6H2Cl2F3N
WDFIJOQQKFWJEA-UHFFFAOYSA-N
3,6-dichloro-2-(trifluoromethyl)pyridine
20213813
97%
Glass Bottle
1g
96% min. (GC)
3,6-dichloro-2-trifluoromethyl pyridine, pyridine, 3,6-dichloro-2-trifluoromethyl, 2,5-dichloro-6-trifluoromethyl pyridine, acmc-20a3gu
C1=CC(=NC(=C1Cl)C(F)(F)F)Cl
215.99
215.99
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3,6-Dichloro-2-(trifluoromethyl)pyridine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Documents