missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)propyl Methanesulfonate, TRC
Click to view available options
Quantity:
2.5 g
5 g
10 g
Unit Size:
10g
2.5g
5g
Specifications
Specifications
| Chemical Name or Material | 3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)propyl Methanesulfonate |
| Analyte or Component Names | 3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)propyl Methanesulfonate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19H21NO5S |
| InChI Formula | InChI=1S/C19H21NO5S/c1-26(22,23)25-12-6-11-20-19(21)24-13-18-16-9-4-2-7-14(16)15-8-3-5-10-17(15)18/h2-5,7-10,18H,6,11-13H2,1H3,(H,20,21) |
| Solution Type | Neat |
| SMILES | O=C(NCCCOS(C)(=O)=O)OCC1C(C=CC=C2)=C2C3=C1C=CC=C3 |
| IUPAC Name | 3-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)propyl methanesulfonate |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 375.44 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction