missing translation for 'onlineSavingsMsg'
Learn More

3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)propyl Methanesulfonate, TRC

Product Code. 30402370
missing translation for 'orderingAttributeHoverText'
Quantity:
2.5 g
5 g
10 g
missing translation for 'unitSize'
10g
2.5g
5g
This item is not returnable. View return policy

Product Code. 30402370

missing translation for 'mfr': Toronto Research Chemicals TRCF6042735G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)propyl Methanesulfonate
Analyte or Component Names 3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)propyl Methanesulfonate
Shipping Condition Room Temperature
Molecular Formula C19H21NO5S
InChI Formula InChI=1S/C19H21NO5S/c1-26(22,23)25-12-6-11-20-19(21)24-13-18-16-9-4-2-7-14(16)15-8-3-5-10-17(15)18/h2-5,7-10,18H,6,11-13H2,1H3,(H,20,21)
Solution Type Neat
SMILES O=C(NCCCOS(C)(=O)=O)OCC1C(C=CC=C2)=C2C3=C1C=CC=C3
IUPAC Name 3-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)propyl methanesulfonate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 375.44
Quantity 5 g
Formula Weight 375.11
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.