missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-(4-tert-butylphenyl)-1,2,4-thiadiazole, 96%, Thermo Scientific™

Catalog Number 15408385
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1086385-86-5
C12H15N3S
233.33
MFCD11655562
KMTRRQKJAQTEEY-UHFFFAOYSA-N
5-4-tert-butylphenyl-1,2,4-thiadiazol-3-amine, 3-amino-5-4-tert-butylphenyl-1,2,4-thiadiazole, 5-4-tert-butyl phenyl-1,2,4-thiadiazol-3-amine
46741383
5-(4-tert-butylphenyl)-1,2,4-thiadiazol-3-amine
CC(C)(C)C1=CC=C(C=C1)C1=NC(N)=NS1
This item is not returnable. View return policy
1086385-86-5
C12H15N3S
233.33
MFCD11655562
KMTRRQKJAQTEEY-UHFFFAOYSA-N
5-4-tert-butylphenyl-1,2,4-thiadiazol-3-amine, 3-amino-5-4-tert-butylphenyl-1,2,4-thiadiazole, 5-4-tert-butyl phenyl-1,2,4-thiadiazol-3-amine
46741383
5-(4-tert-butylphenyl)-1,2,4-thiadiazol-3-amine
CC(C)(C)C1=CC=C(C=C1)C1=NC(N)=NS1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 221°C to 222°C
Quantity 250 mg
Formula Weight 233.33
Percent Purity 96%
Chemical Name or Material 3-Amino-5-(4-tert-butylphenyl)-1,2,4-thiadiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'