missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminoisoxazole-4-carbonitrile, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 3-Aminoisoxazole-4-carbonitrile |
| CAS | 258518-65-9 |
| Molecular Formula | C4 H3 N3 O |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C4H3N3O/c5-1-3-2-8-7-4(3)6/h2H,(H2,6,7) |
| SMILES | N=C1NOC=C1C#N |
| IUPAC Name | 3-imino-1,2-oxazole-4-carbonitrile |
| Molecular Weight (g/mol) | 109.086 |
| Formula Weight | 109.028 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction