missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ 3-AP

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
3-AP is a ribonucleotide reductase inhibitor. Also an iron chelator. Inhibits leukemia cell growth and acts in synergy with cisplatin (Cat. No.2251). Active in vivo.
Specifications
Specifications
| Chemical Name or Material | 2-[(3-Amino-2-pyridinyl)methylene]hydrazinecarbothioamide |
| CAS | 143621-35-6 |
| Synonym | 3-aminopyridine-2-carboxaldehyde thiosemicarbazone |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in DMSO |
| Molecular Formula | C7H9N5S |
| Formula Weight | 195.24 |
| Physical Form | Solid |
| Color | White |
| Product Type | Bioactive Small Molecule |
| Show More |
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction