missing translation for 'onlineSavingsMsg'
Learn More

3-Bromoanisole, 99+%

Product Code. p-71596 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
This item is not returnable. View return policy

Product Code. 10559980

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2398-37-0 | C7H7BrO | 187.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2398-37-0
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.04
MDL Number MFCD00000081
InChI Key PLDWAJLZAAHOGG-UHFFFAOYSA-N
Synonym 3-bromoanisole, m-bromoanisole, 3-methoxybromobenzene, 3-bromo anisole, m-bromomethoxybenzene, benzene, 1-bromo-3-methoxy, anisole, m-bromo, 3-methoxy-1-bromobenzene, m-methoxybromobenzene, meta-bromoanisole
PubChem CID 16971
IUPAC Name 1-bromo-3-methoxybenzene
SMILES COC1=CC=CC(Br)=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 2.0°C
Color Brown-Yellow to Colorless
Density 1.4700g/mL
Boiling Point 210.0°C to 211.0°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4OCH3
Refractive Index 1.5630 to 1.5650
Quantity 25 mL
Beilstein 06, 198
Specific Gravity 1.47
Formula Weight 187.04
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 3-Bromoanisole, 99+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.