missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(Bromomethyl)thiophene, 97%

Brand:  Alfa Aesar™ H61497.06

Additional Details : CAS : 34846-44-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15408058

  • 382.00€ / 5g


Estimated Shipment
11-05-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

34846-44-1
177.059
KBWHYRUAHXHHFO-UHFFFAOYSA-N
520703
C1=CSC=C1CBr
C5H5BrS
MFCD03840497
3-bromomethyl thiophene, 3-bromomethyl-thiophene, 3-thenyl bromide, thiophene, 3-bromomethyl, 3-thienylmethyl bromide, thien-3-ylmethyl bromide, zlchem 651, 3-brommethyl-thiophen, pubchem5295, 3 bromomethyl thiophene
3-(bromomethyl)thiophene

Specifications

3-(Bromomethyl)thiophene
1.603
76°C (1mmHg)
5g
C5H5BrS
106459
KBWHYRUAHXHHFO-UHFFFAOYSA-N
3-(bromomethyl)thiophene
520703
Liquid
1.616 g/mL
Moisture sensitive
Brown
34846-44-1
MFCD03840497
3-bromomethyl thiophene, 3-bromomethyl-thiophene, 3-thenyl bromide, thiophene, 3-bromomethyl, 3-thienylmethyl bromide, thien-3-ylmethyl bromide, zlchem 651, 3-brommethyl-thiophen, pubchem5295, 3 bromomethyl thiophene
C1=CSC=C1CBr
177.059
177.06
97%
Videos
SDS Documents