missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromothiophene, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
3-Bromothiophene is used in the preparation of derivatives such as thienylenic alpha, μ-diformyl-alpha-oligothiophenes and 3-lithiothiophene. It is also used in the synthesis of 3-lithiothiophene by reacting with n-butyllithium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Bromothiophene is used in the preparation of derivatives such as thienylenic alpha, µ-diformyl-alpha-oligothiophenes and 3-lithiothiophene. It is also used in the synthesis of 3-lithiothiophene by reacting with n-butyllithium.
Solubility
Immiscible with water.
Notes
Light sensitive. Incompatible with strong oxidizing agents, strong reducing agents and strong bases.
3-Bromothiophene is used in the preparation of derivatives such as thienylenic alpha, µ-diformyl-alpha-oligothiophenes and 3-lithiothiophene. It is also used in the synthesis of 3-lithiothiophene by reacting with n-butyllithium.
Solubility
Immiscible with water.
Notes
Light sensitive. Incompatible with strong oxidizing agents, strong reducing agents and strong bases.
Specifications
Density | 1.722 |
Boiling Point | 157°C to 158°C |
Flash Point | 63°C (145°F) |
Odor | Pungent |
Refractive Index | 1.591 |
Quantity | 10 g |
UN Number | UN2810 |
Beilstein | 105338 |
Solubility Information | Immiscible with water. |
Formula Weight | 163.04 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'