Learn More
3-Chloro-2-methoxypyridine-4-boronic acid, 95%, ACROS Organics™
Brand: Acros Organics 448770010
Additional Details : CAS : 957060-88-7 Poids : 0.01000kg
Packaging | Glass bottle |
---|---|
Quantity | 1g |
Chemical Identifiers
957060-88-7 | |
187.39 | |
3-chloro-2-methoxypyridine-4-boronic acid, 3-chloro-2-methoxypyridin-4-yl boronic acid, 3-chloro-2-methoxypyridin-4-ylboronicacid, 2-methoxy-3-chloropyridine-4-boronic acid, 3-chloro-2-methoxy-4-pyridineboronic acid, pubchem17058, abbypharma ap-17-5909, 3-chloro-2-methoxypyrdine-4-boronic acid, boronic acid, b-3-chloro-2-methoxy-4-pyridinyl | |
(3-chloro-2-methoxypyridin-4-yl)boronic acid |
C6H7BClNO3 | |
YQNAXOGPSUVHNU-UHFFFAOYSA-N | |
44119809 | |
B(C1=C(C(=NC=C1)OC)Cl)(O)O |
Specifications
3-Chloro-2-methoxypyridine-4-boronic acid | |
140°C | |
957060-88-7 | |
C6H7BClNO3 | |
YQNAXOGPSUVHNU-UHFFFAOYSA-N | |
(3-chloro-2-methoxypyridin-4-yl)boronic acid | |
44119809 | |
95% |
Glass bottle | |
1g | |
94% min. (HPLC) | |
3-chloro-2-methoxypyridine-4-boronic acid, 3-chloro-2-methoxypyridin-4-yl boronic acid, 3-chloro-2-methoxypyridin-4-ylboronicacid, 2-methoxy-3-chloropyridine-4-boronic acid, 3-chloro-2-methoxy-4-pyridineboronic acid, pubchem17058, abbypharma ap-17-5909, 3-chloro-2-methoxypyrdine-4-boronic acid, boronic acid, b-3-chloro-2-methoxy-4-pyridinyl | |
B(C1=C(C(=NC=C1)OC)Cl)(O)O | |
187.39 | |
187.39 |
Safety and Handling
- 3-Chloro-2-methoxypyridine-4-boronic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.