missing translation for 'onlineSavingsMsg'
Learn More

(3-Chloropropyl)trichlorosilane, 97%, Thermo Scientific™

Product Code. 11375129
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11375129

Brand: Thermo Scientific Alfa Aesar A17770.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2550-06-3
MDL Number MFCD00013694

Specifications

Boiling Point 181°C to 183°C
Quantity 100 g
Molecular Formula C12H24O4Si4
Linear Formula Cl(CH2)3SiCl3
UN Number UN2987
Beilstein 1737851
Flash Point 84°C (183°F)
Synonym 2,4,6,8-tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane, 2,4,6,8-tetramethyl-2,4,6,8-tetravinyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane, cyclotetrasiloxane, 2,4,6,8-tetraethenyl-2,4,6,8-tetramethyl, 1,3,5,7-tetravinyl-1,3,5,7-tetramethylcyclotetrasiloxane, 1,3,5,7-tetramethyl-1,3,5,7-tetravinylcyclotetrasiloxane, 2,4,6,8-tetravinyl-2,4,6,8-tetramethylcyclotetrasiloxane, tetravinyltetramethylcyclotetrasiloxane, s-tetramethyltetravinylcyclotetrasiloxane, unii-3nt606t1vk, cyclotetrasiloxane, tetraethenyltetramethyl
InChI Key VMAWODUEPLAHOE-UHFFFAOYSA-N
SMILES C[Si]1(O[Si](O[Si](O[Si](O1)(C)C=C)(C)C=C)(C)C=C)C=C
IUPAC Name 2,4,6,8-tetrakis(ethenyl)-2,4,6,8-tetramethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
Molecular Weight (g/mol) 344.66
PubChem CID 75706
Formula Weight 211.98
Percent Purity 97%
Odor Acrid
Refractive Index 1.4675
Sensitivity Moisture sensitive
Density 1.36
Chemical Name or Material (3-Chloropropyl)trichlorosilane
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.