missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanobenzyl alcohol, 97%, Thermo Scientific Chemicals

Catalog Number 15410558
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
874-97-5
C8H7NO
133.15
MFCD01321087
PCOFIIVWHXIDGT-UHFFFAOYSA-N
3-cyanobenzyl alcohol, 3-hydroxymethyl benzonitrile, 3-cyanobenzylalcohol, benzonitrile, 3-hydroxymethyl, 3-hyroxymethylbenzonitrile, 3-hydroxymethyl benzenecarbonitrile, m-cyanobenzyl alcohol, 3-cyanophenyl methanol, acmc-209qo5
2734677
3-(hydroxymethyl)benzonitrile
C1=CC(=CC(=C1)CO)C#N
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
874-97-5
C8H7NO
133.15
MFCD01321087
PCOFIIVWHXIDGT-UHFFFAOYSA-N
3-cyanobenzyl alcohol, 3-hydroxymethyl benzonitrile, 3-cyanobenzylalcohol, benzonitrile, 3-hydroxymethyl, 3-hyroxymethylbenzonitrile, 3-hydroxymethyl benzenecarbonitrile, m-cyanobenzyl alcohol, 3-cyanophenyl methanol, acmc-209qo5
2734677
3-(hydroxymethyl)benzonitrile
C1=CC(=CC(=C1)CO)C#N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Flash Point 109°C (228°F)
Quantity 1 g
Formula Weight 133.15
Percent Purity 97%
Chemical Name or Material 3-Cyanobenzyl alcohol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'