missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanopyridine-5-boronic acid pinacol ester, 96%

Product Code. 15470688
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15470688

Brand: Thermo Scientific Alfa Aesar H54385.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 402718-29-0 | C12H15BN2O2 | 230.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 402718-29-0
Molecular Formula C12H15BN2O2
Molecular Weight (g/mol) 230.07
MDL Number MFCD07780755
InChI Key BOIKCRIMIQAFQJ-UHFFFAOYSA-N
Synonym 3-cyanopyridine-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl nicotinonitrile, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carbonitrile, nicotinonitril-5-boronic acid, 5-cyanopyridine-3-boronic acid pinacol ester, nicotinonitrile-5-boronic acid pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carbonitrile, 3-cyano-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl nicotinonitrile?, 3-cyano-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine
PubChem CID 16414216
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carbonitrile
SMILES CC1(C)OB(OC1(C)C)C1=CC(=CN=C1)C#N

Specifications

Melting Point 122°C to 124°C
Quantity 1 g
Formula Weight 230.07
Percent Purity 96%
Chemical Name or Material 3-Cyanopyridine-5-boronic acid pinacol ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.