Learn More
3-Deoxy-D-glucosone, 95%, Thermo Scientific Chemicals
Catalog Number 15407499
Click to view available options
Quantity:
1 mg
5 mg
4084-27-9
C6H10O5
162.141
MFCD01723101
ZGCHLOWZNKRZSN-NTSWFWBYSA-N
3-deoxyglucosone, 3-deoxy-d-glucosone, 3-deoxy-d-erythro-hexos-2-ulose, d-3-deoxyglucosone, 3-deoxy-d-erythro-hexosulose, 3-deoxy-d-fructose, ccris 4272, 3-deoxy-2-ketoglucose, d-erythro-hexos-2-ulose, 3-deoxy, unii-exv5374vey
114839
CHEBI:60777
(4S,5R)-4,5,6-trihydroxy-2-oxohexanal
C(C(C(CO)O)O)C(=O)C=O
4084-27-9
C6H10O5
162.141
MFCD01723101
ZGCHLOWZNKRZSN-NTSWFWBYSA-N
3-deoxyglucosone, 3-deoxy-d-glucosone, 3-deoxy-d-erythro-hexos-2-ulose, d-3-deoxyglucosone, 3-deoxy-d-erythro-hexosulose, 3-deoxy-d-fructose, ccris 4272, 3-deoxy-2-ketoglucose, d-erythro-hexos-2-ulose, 3-deoxy, unii-exv5374vey
114839
CHEBI:60777
(4S,5R)-4,5,6-trihydroxy-2-oxohexanal
C(C(C(CO)O)O)C(=O)C=O
Frequently used as a reference for the analysis and detection of glucose degradation products and glycating agents. Also used for cross-linking studies as it readily reacts with protein amino groups.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Frequently used as a reference for the analysis and detection of glucose degradation products and glycating agents
  • Also used for cross-linking studies as it readily reacts with protein amino groups
TRUSTED_SUSTAINABILITY
Color Yellow
Quantity 1 mg
Solubility Information Soluble in water,methanol or hot ethanol.
Formula Weight 162.14
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-Deoxy-D-glucosone