Learn More
BRAND™ 3-Device Charging Stand
Shop All Brand Products
Click to view available options
Plug Type:
USA/Japan
Unit Size:
Each
Description
- Securely holds and charges 3 Transferpette™ electronic instruments
- PRI 100-240 V AC adapter
- Choose UK/Ireland or USA/Japan plug type
Specifications
Specifications
| Type | Charging Stand, 3 Device |
| For Use With (Equipment) | Transferpette™ electronic |
| Plug Type | USA/Japan |
| Quantity | 1 Each |
| Electrical Requirements | 230V, 50Hz |
| Voltage | 230 V |
| Frequency | 50 Hz |