missing translation for 'onlineSavingsMsg'
Learn More

3-Dimethylamino-2-methylpropyl Chloride Hydrochloride, TRC

Product Code. 30381966
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 30381966

Brand: Toronto Research Chemicals TRCD47079550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Dimethylamino-2-methylpropyl Chloride Hydrochloride
CAS 4261-67-0
Molecular Formula C6H15Cl2N
Quantity 50 mg
Synonym 3-Chloro-N,N,2-trimethyl-1-propanamine Hydrochloride, 1-Chloro-2-methyl-3-(dimethylamino)propane Hydrochloride, 1-Chloro-3-(N,N-dimethylamino)-2-methylpropane Hydrochloride, 2,N,N-Trimethylaminopropyl Chloride Hydrochloride
InChI Formula InChI=1S/C6H14ClN.ClH/c1-6(4-7)5-8(2)3;/h6H,4-5H2,1-3H3;1H
SMILES Cl.CC(CCl)CN(C)C
Molecular Weight (g/mol) 172.1
Formula Weight 171.0582
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.