missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxy-2,6-difluorophenylacetic acid, 97%, Thermo Scientific™

Catalog Number 15415627
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1092461-32-9
C10H10F2O3
216.184
MFCD11519374
CXLCGFWZQHJZNB-UHFFFAOYSA-N
3-ethoxy-2,6-difluorophenylacetic acid, 3-ethoxy-2,6-difluorophenyl acetic acid, 2-3-ethoxy-2,6-difluorophenyl acetic acid, 3-ethoxy-2,6-difluorophenylacetic acid, jrd
18444711
2-(3-ethoxy-2,6-difluorophenyl)acetic acid
CCOC1=C(C(=C(C=C1)F)CC(=O)O)F
This item is not returnable. View return policy
1092461-32-9
C10H10F2O3
216.184
MFCD11519374
CXLCGFWZQHJZNB-UHFFFAOYSA-N
3-ethoxy-2,6-difluorophenylacetic acid, 3-ethoxy-2,6-difluorophenyl acetic acid, 2-3-ethoxy-2,6-difluorophenyl acetic acid, 3-ethoxy-2,6-difluorophenylacetic acid, jrd
18444711
2-(3-ethoxy-2,6-difluorophenyl)acetic acid
CCOC1=C(C(=C(C=C1)F)CC(=O)O)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 101°C to 103°C
Quantity 250 mg
Formula Weight 216.18
Percent Purity 97%
Chemical Name or Material 3-Ethoxy-2,6-difluorophenylacetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'