missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-1-pentyn-3-ol, 98%, Thermo Scientific Chemicals

Catalog Number p-7058310
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
6285-06-9
MFCD00014392
CHEBI:52687
This item is not returnable. View return policy
6285-06-9
MFCD00014392
CHEBI:52687

3-Ethyl-1-pentyn-3-ol is used in agrochemical, pharmaceutical and dyestuff field .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Ethyl-1-pentyn-3-ol is used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 0.873
Boiling Point 138°C to 140°C
Flash Point 38°C (100°F)
Molecular Formula C4H10O2
Refractive Index 1.438
Quantity 5 g
UN Number UN1987
Beilstein 1740944
Synonym r---1,3-butanediol, r-butane-1,3-diol, 3r-butane-1,3-diol, r-1,3-butanediol, 1,3-butanediol, 3r, r---1,3-butylene glycol, r---butane-1,3-diol, 1,3-butanediol, r
Solubility Information Not miscible in water.
InChI Key PUPZLCDOIYMWBV-UHFFFAOYNA-N
SMILES CC(CCO)O
IUPAC Name (3R)-butane-1,3-diol
Molecular Weight (g/mol) 90.12
PubChem CID 637497
Formula Weight 112.17
Percent Purity 98%
Chemical Name or Material 3-Ethyl-1-pentyn-3-ol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'