missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-(hydroxymethyl)pyridine hydrochloride, 85%, technical, Thermo Scientific™

Catalog Number p-7077149
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
14173-30-9
C6H7NO2·HCl
161.59
SKVRYQUMKJQZFN-UHFFFAOYSA-N
3-hydroxy-2-pyridinemethanol hydrochloride, 2-hydroxymethyl pyridin-3-ol hydrochloride, 3-hydroxy-2-hydroxymethyl pyridine hydrochloride, 2-pyridinemethanol, 3-hydroxy-, hydrochloride, 2-hydroxymethyl pyridin-3-ol hcl, 3-hydroxy-2-hydroxymethylpyridine hydrochloride, pubchem4075, 3-hydroxy-2-hydroxymethyl pyridine hcl, acmc-209xde, 3-hydroxy-2-pyridinemethanol hcl
3084185
2-(hydroxymethyl)pyridin-3-ol;hydrochloride
C1=CC(=C(N=C1)CO)O.Cl
This item is not returnable. View return policy
14173-30-9
C6H7NO2·HCl
161.59
SKVRYQUMKJQZFN-UHFFFAOYSA-N
3-hydroxy-2-pyridinemethanol hydrochloride, 2-hydroxymethyl pyridin-3-ol hydrochloride, 3-hydroxy-2-hydroxymethyl pyridine hydrochloride, 2-pyridinemethanol, 3-hydroxy-, hydrochloride, 2-hydroxymethyl pyridin-3-ol hcl, 3-hydroxy-2-hydroxymethylpyridine hydrochloride, pubchem4075, 3-hydroxy-2-hydroxymethyl pyridine hcl, acmc-209xde, 3-hydroxy-2-pyridinemethanol hcl
3084185
2-(hydroxymethyl)pyridin-3-ol;hydrochloride
C1=CC(=C(N=C1)CO)O.Cl
Melting Point 217.0°C
Infrared Spectrum Authentic
Assay Percent Range 84% min. (HPLC)
Linear Formula HO(C5H3N)CH2OH
Quantity 5 g
Formula Weight 161.59
Percent Purity 85%
Grade Technical
Chemical Name or Material 3-Hydroxy-2-(hydroxymethyl)pyridine hydrochloride
compliance-icons
Product Identifier
  • 3-Hydroxy-2-(hydroxymethyl)pyridine hydrochloride, 85%, Technical
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'