missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-2-propynyl N-Butylcarbamate, TRC

Product Code. 30136744
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
Unit Size:
1g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136744 50 g 50g
30132397 5 g 5g
18014020 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136744 Supplier Toronto Research Chemicals Supplier No. TRCI71895050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55406-53-6
InChI Formula InChI=1S/C8H12INO2/c1-2-3-6-10-8(11)12-7-4-5-9/h2-3,6-7H2,1H3,(H,10,11)
IUPAC Name 3-iodoprop-2-ynyl N-butylcarbamate
Molecular Formula C8 H12 I N O2
Molecular Weight (g/mol) 281.09
Chemical Name or Material Iodocarb (IPBC)

Specifications

Quantity 50 g
Synonym Carbamic acid, butyl-, 3-iodo-2-propynyl ester (9CI), 3-Iodo-2-propynyl N-butylcarbamate, 3-Iodo-2-propynyl butylcarbamate, 3-Iodopropargyl n-butylcarbamate, Acticide IPS 20, Acticide IPW 50, Bioban I 20, Biodocarb, Biodocarb L 15BDG, Biodocarb L 20TEX, Biodocarb L 30BDG, Coatcide 123, Dekaben LMB, Denicide XR 5, Ecobois, FL 10, Fungitrol 400, Fungitrol 720, Fungitrol 820, Fungitrol 920, Fungitrol 940, Fungitrol F 10, Glycacil, Glycacil 2000, Glycasil L, Guardsan 388, Hokucide IPB, IBF 8DOP, IPBC, IPBC 20, IPBC 40, Iodocarb, Iodopropynyl butylcarbamate, MP 100, Mergal 710S, N-Butyl 3-iodo-2-propyn-1-yl carbamate, Neomid 220, Nipacide IPBC 20, Omacide IPBC, Omacide IPBC 100, P 20T, Polyphase, Polyphase 60B, Polyphase 641, Polyphase AF 1, Polyphase CST, Polyphase P 20T, Polyphase P-20T, Polyphase P100, Preventol MP 100, Preventol MP 260, Preventol MP 400, Thompson's Wood Protector, Timbertreat D, Troysan P 20T, Troysan Polyphase 60B, Troysan Polyphase AF 1, Troysan Polyphase AF 3, Troysan Polyphase P 100, Troysan Polyphase P 20T, Troysan Polyphase WD 17, Yotoru IP
SMILES CCCCNC(=O)OCC#CI
Formula Weight 280.9913
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.