Learn More
Alfa Aesar™ 3'-Methoxybiphenyl-4-sulfonyl chloride, 96%
Brand: Alfa Aesar™ H33211.03
Additional Details : CAS : 186550-26-5 Weight : 0.00100kg Transport : UN number : 3261 Chem class : 8 Pack group : III
Quantity | 1g |
---|
Chemical Identifiers
186550-26-5 | |
282.738 | |
DVQBUJTYQXKLKZ-UHFFFAOYSA-N | |
10826798 | |
COC1=CC=CC(=C1)C2=CC=C(C=C2)S(=O)(=O)Cl |
C13H11ClO3S | |
MFCD06409270 | |
3'-methoxy-biphenyl-4-sulfonyl chloride, 3'-methoxybiphenyl-4-sulfonyl chloride, 3'-methoxy 1,1'-biphenyl-4-sulfonyl chloride, 3'-methoxy-1,1'-biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulfonylchloride, 3'-methoxy, 4-3-methoxyphenyl benzenesulfonyl chloride, 3'-methoxy-biphenyl-4-sulfonylchloride, 3/'-methoxybiphenyl-4-sulfonyl chloride | |
4-(3-methoxyphenyl)benzenesulfonyl chloride |
Specifications
3'-Methoxybiphenyl-4-sulfonyl chloride | |
80°C to 84°C | |
186550-26-5 | |
MFCD06409270 | |
3'-methoxy-biphenyl-4-sulfonyl chloride, 3'-methoxybiphenyl-4-sulfonyl chloride, 3'-methoxy 1,1'-biphenyl-4-sulfonyl chloride, 3'-methoxy-1,1'-biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulfonylchloride, 3'-methoxy, 4-3-methoxyphenyl benzenesulfonyl chloride, 3'-methoxy-biphenyl-4-sulfonylchloride, 3/'-methoxybiphenyl-4-sulfonyl chloride | |
COC1=CC=CC(=C1)C2=CC=C(C=C2)S(=O)(=O)Cl | |
282.738 | |
282.74 |
Moisture sensitive | |
1g | |
C13H11ClO3S | |
UN3261 | |
DVQBUJTYQXKLKZ-UHFFFAOYSA-N | |
4-(3-methoxyphenyl)benzenesulfonyl chloride | |
10826798 | |
96% |