Learn More
3-Methylstyrene, 95%, Thermo Scientific Chemicals
Catalog Number p-7077353
Click to view available options
Quantity:
1 g
5 g
100-80-1
C9H10
118.18
JZHGRUMIRATHIU-UHFFFAOYSA-N
3-methylstyrene, 3-vinyltoluene, 1-methyl-3-vinylbenzene, m-vinyltoluene, m-methylstyrene, benzene, 1-ethenyl-3-methyl, styrene, m-methyl, m-methyl styrene, meta-methylstyrene, unii-ffk4i9430g
7529
1-ethenyl-3-methylbenzene
CC1=CC=CC(=C1)C=C
100-80-1
C9H10
118.18
JZHGRUMIRATHIU-UHFFFAOYSA-N
3-methylstyrene, 3-vinyltoluene, 1-methyl-3-vinylbenzene, m-vinyltoluene, m-methylstyrene, benzene, 1-ethenyl-3-methyl, styrene, m-methyl, m-methyl styrene, meta-methylstyrene, unii-ffk4i9430g
7529
1-ethenyl-3-methylbenzene
CC1=CC=CC(=C1)C=C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point -82°C to 81°C
Boiling Point 170.0°C to 171.0°C
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 118.18
Percent Purity 95%
Chemical Name or Material 3-Methylstyrene
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Methylstyrene, 98%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.