missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Morpholinomethyl)benzaldehyde, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Assay Percent Range | 95% |
Quantity | 1 g |
Formula Weight | 205.26 |
Percent Purity | 95% |
Chemical Name or Material | 3-(Morpholinomethyl)benzaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'