missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrophthalic Acid, TRC

Product Code. 30470307
Change view
Click to view available options
Quantity:
10 g
250 g
Unit Size:
250g
Product Code. Quantity unitSize
30470307 250 g 250g
30473294 10 g -
2 options
This item is not returnable. View return policy

Product Code. 30470307

Brand: Toronto Research Chemicals TRCN515295250G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-11-2
Molecular Formula C8 H5 N O6
Molecular Weight (g/mol) 211.13
Synonym 1,2-Benzenedicarboxylic acid, 3-nitro- (9CI, ACI), 3-Nitro-1,2-benzenedicarboxylic acid (ACI), Phthalic acid, 3-nitro- (6CI, 7CI, 8CI), 3-Nitrophthalic acid, m-Nitrophthalic acid, NSC 3120, o-Nitrophthalic acid
IUPAC Name 3-nitrophthalic acid
SMILES OC(=O)c1cccc(c1C(=O)O)[N+](=O)[O-]

Specifications

Quantity 250 g
InChI Formula InChI=1S/C8H5NO6/c10-7(11)4-2-1-3-5(9(14)15)6(4)8(12)13/h1-3H,(H,10,11)(H,12,13)
Formula Weight 211.0117
Chemical Name or Material 3-Nitrophthalic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.