Learn More
3-Part Latex Free Insulin Syringes
Use these syringes for the aspiration of fluids or sampling blood or other body fluids. Terumo™ 3-Part Latex Free Insulin Syringes feature a unique double beveled design for greater patient comfort.
Brand:  Terumo 8SS01H1
Additional Details : Weight : 0.34020kg
Product Code. 11750425
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • Ultra-sharp needles
  • Contain no components made of natural rubber or latex
  • Clear barrels
  • Bold, precise scale markings
  • Smooth plunger action
  • Tamper evident peel packs
Specifications
Specifications
Yes
Insulin Syringe
100/Pk.
1 mL