missing translation for 'onlineSavingsMsg'
Learn More

3-Penten-2-one, tech. 85%, Thermo Scientific Chemicals

Catalog Number p-7046074
24.60 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
625-33-2
C5H8O
84.118
MFCD00009290
LABTWGUMFABVFG-ONEGZZNKSA-N
z-pent-3-en-2-one, unii-w8xx21whyc, z-3-penten-2-one, w8xx21whyc, 3-penten-2-one, 3z-pent-3-en-2-one, 3-penten-2-one, z, cis-3-penten-2-one, 3-penten-2-one, cis
637920
(E)-pent-3-en-2-one
CC=CC(=O)C
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
625-33-2
C5H8O
84.118
MFCD00009290
LABTWGUMFABVFG-ONEGZZNKSA-N
z-pent-3-en-2-one, unii-w8xx21whyc, z-3-penten-2-one, w8xx21whyc, 3-penten-2-one, 3z-pent-3-en-2-one, 3-penten-2-one, z, cis-3-penten-2-one, 3-penten-2-one, cis
637920
(E)-pent-3-en-2-one
CC=CC(=O)C

3-Penten-2-one

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Penten-2-one

Solubility
Slightly soluble in Water

Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with bases, reducing agents and oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 0.862
Boiling Point 119°C to 121°C
Flash Point 21°C (69°F)
Odor Pungent, Makes Eyes Water
Refractive Index 1.4355
Quantity 1 g
UN Number UN1992
Beilstein 1633505
Solubility Information Slightly soluble in Water
Formula Weight 84.12
Percent Purity 85%
Grade Technical
Chemical Name or Material 3-Penten-2-one
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'