missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrimidin-5-ylaniline, ≥97%, Thermo Scientific™

Catalog Number p-3176068
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
69491-59-4
C10H9N3
171.203
MFCD06802522
DZEIKJMNXHOFHL-UHFFFAOYSA-N
7162049
3-pyrimidin-5-ylaniline
C1=CC(=CC(=C1)N)C2=CN=CN=C2
This item is not returnable. View return policy
69491-59-4
C10H9N3
171.203
MFCD06802522
DZEIKJMNXHOFHL-UHFFFAOYSA-N
7162049
3-pyrimidin-5-ylaniline
C1=CC(=CC(=C1)N)C2=CN=CN=C2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Synonym 3-pyrimidin-5-yl aniline, 3-5-pyrimidinyl aniline, benzenamine,3-5-pyrimidinyl, 3-5-pyrimidyl aniline, 3-5-pyrimidyl-aniline, 3-pyrimidin-5-ylphenylamine, 3-pyrimidin-5-yl-phenylamine, 3-pyrimidin-5-yl phenylamine, benzenamine, 3-5-pyrimidinyl
Formula Weight 171.2
Percent Purity 97%
Chemical Name or Material 3-Pyrimidin-5-ylaniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'