missing translation for 'onlineSavingsMsg'
Learn More

3-Quinuclidinol, Mikromol

Product Code. 30459210
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30459210

Brand: Mikromol MM0825.02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3-Quinuclidinol, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Quinuclidinol
CAS 1619-34-7
Analyte or Component Names 3-Quinuclidinol
Shipping Condition Room Temperature
Molecular Formula C7 H13 N O
Certifications/Compliance ISO 17025
Synonym 3-Quinuclidinol
InChI Formula InChI=1S/C7H13NO/c9-7-5-8-3-1-6(7)2-4-8/h6-7,9H,1-5H2
Solution Type Neat
SMILES OC1CN2CCC1CC2
IUPAC Name 1-azabicyclo[2.2.2]octan-3-ol
Molecular Weight (g/mol) 127.18
Quantity 100 mg
Formula Weight 127.0997 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.