missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Quinuclidinol, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 3-Quinuclidinol |
| CAS | 1619-34-7 |
| Molecular Formula | C7 H13 N O |
| Quantity | 5 g |
| Synonym | 3-Quinuclidinol, 1-Azabicyclo[2.2.2]octan-3-ol, (±)-1-Azabicyclo[2.2.2]octan-3-ol, 3-Quinuclidinol (6CI,7CI,8CI), (RS)-3-Quinuclidinol, (±)-3-Quinuclidinol, 1-Azabicyclo[2.2.2]oct-3-yl alcohol, 3-Hydroxy-1-azabicyclo[2.2.2]octane, 3-Hydroxyquinuclidine, 4-Azabicyclo[2.2.2]octan-2-ol, NSC 93905, dl-3-Quinuclidinol |
| InChI Formula | InChI=1S/C7H13NO/c9-7-5-8-3-1-6(7)2-4-8/h6-7,9H,1-5H2 |
| SMILES | OC1CN2CCC1CC2 |
| IUPAC Name | 1-azabicyclo[2.2.2]octan-3-ol |
| Molecular Weight (g/mol) | 127.18 |
| Formula Weight | 127.0997 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction