Learn More
Cole-Parmer™ 304 Stainless Steel Cover
Shop All Cole-Parmer Products
Click to view available options
Diameter (Metric) Outer:
171.45 mm
222mm
240mm
280mm
Unit Size:
Each
Description
- Covers reduce evaporation
- Contain harmful vapors, or keep contaminants out
Specifications
Specifications
| Diameter (Metric) Outer | 280mm |
| Material | Stainless Steel (304) |
| Quantity | 1Each |
| Certifications/Compliance | AISI 304 |