missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 384-Well, Poly-D Lysine-Treated, Flat-Bottom Microplate

Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid

Brand:  Corning™ 3768

Additional Details : Weight : 22.40000kg

Product Code. 15585283

  • 3515.62€ / Pack of 100
Estimated Shipment: 05-06-2024
to see stock.
Add to basket

Description

Description

Suited for fluorescent and luminescent assays using either top or bottom detection microplate readers.

Specifications

Specifications

Standard
0.06 cm2
Black, Clear Bottom
384
Poly-D-Lysine
100/Cs.
127.8 x 85.5 x 14.35 mm (5 x 3.36 x 0.56 in.)
Top/bottom: 3.63/2.82 mm
With Lid
16 x 24
Polystyrene
Sterile
Flat Bottom (F-well)
20/Pk.
11.6 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions