Learn More
Greiner Bio-One 384-well Polypropylene Microplates
Manufactured with polypropylene for optimum chemical resistance and high temperature tolerance. Greiner Bio-One™ 384 Well Polypropylene Microplates provide four times the number of wells in a 96 well standard microplate while retaining external dimensions compatible with standard equipment and automated systems.
Brand: Greiner Bio-One 781280
Additional Details : Weight : 4.16700kg
Description
- Footprint compatible with automated systems
- Can be traced to production through defined LOT number system
- Free of detectable DNase, RNase and human DNA
- Non-pyrogenic
- Manufactured under DIN ISO 9001 guidelines
- Well-to-well spacing: 4.5mm
- Low biomolecular binding characteristics
- Resistant to many standard laboratory chemicals, such as DMSO
Specifications
DIN ISO 9001 | |
Standard | |
Inner packs of 10 | |
Genomic | |
384 | |
Non-sterile | |
Conical Bottom (V-well) |
DNase/RNase and human DNA free | |
16 x 24 | |
Natural | |
Polypropylene | |
10 to 130 μL | |
138 μL | |
100/Cs. |