Learn More
Greiner Bio-One 384-well Polystyrene Non-binding Flat Bottom Microplate
GREENER_CHOICE
Product Code. 11866091
Click to view available options
Color:
Black
White
Unit Size:
Pack of 40
Product Code. 11866091
Brand: Greiner Bio-One 781900
Currently Unavailable: We have no information when this product will be available.
View alternative products
Provides an ultra low non-specific biomolecular binding properties. Greiner Bio-One 384 Well Polystyrene Non-binding Flat Bottom Microplate has long-term surface stability without degradation or leaching.
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Higher assay sensitivity with reduced background
Service Plan
No
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 16 x 24
Color Black
Material Polystyrene
No. of Wells 384
Sterility Non-sterile
Well Shape Flat Bottom (F-well)