Learn More
Greiner Bio-One 384-well Polystyrene Non-binding Flat Bottom Microplate
Click to view available options
Color:
Black
White
Unit Size:
Pack of 40
Description
- Free of detectable DNase, RNase, human DNA
- Non-pyrogenic
- Manufactured under DIN ISO 9001 guidelines
- Higher assay sensitivity with reduced background
Service Plan
No
Specifications
Specifications
| Format | Standard |
| Array | 16 x 24 |
| Autoclavable | Not Autoclavable |
| Binding Type | Non Binding |
| Color | White |
| Product Type | Microplate |
| Material | Polystyrene |
| No. of Wells | 384 |
| Volume (Metric) Working | 15 to 110 μL |
| Sterility | Non-sterile |
| Show More |