Learn More
Greiner Bio-One 384-well Polystyrene Streptavidin-coated Flat Bottom Microplate
GREENER_CHOICE
Product Code. 11985315
Click to view available options
Color:
Black
Black, Clear Bottom
Clear
White
Unit Size:
Pack of 40
Pack of 5
Product Code. 11985315
Brand: Greiner Bio-One 781995
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Pre-blocked and ready-to-use.
  • Manufactured under DIN ISO 9001 guidelines
  • Biotin binding capacity: >1.5ng/well or >6pmol/well
  • Solid bottom
  • Storable at room temperature
  • With lid
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 16 x 24
Autoclavable Not Autoclavable
Color White
Material Polystyrene
No. of Wells 384
Sterility Non-sterile
Surface Treatment Streptavidin
Well Shape Rounded Square
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 40
No. per Pack 5
Temperature Range (Metric) Room Temperature