Learn More
BRAND™ 384-Well Skirted Microplates, BIO-CERT™ PCR QUALITY
Product Code. 10751193 Shop All Brand Products
Change view
Click to view available options
Color:
Clear
White
Unit Size:
Pack of 50
Product Code. Color unitSize
10751193 Clear Pack of 50
10440095 White Pack of 50
2 options
Product Code. 10751193
Brand: BRAND™ 781345
Only null left
Add to basket
Add to basket
Optimized for reliable and precise results in a wide range of PCR applications.
BRAND™ PCR products deliver excellent, reproducible results for a wide range of PCR and qPCR applications. Manufactured from high-quality virgin resins, these products are free from lubricants, chemical additives, DNA, DNase, RNase, and other PCR inhibitors. Key features include:
  • Exceptional Quality and Purity
    • BIO-CERT™ PCR QUALITY ensures plates are free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors
    • Free of contaminants that could interfere with reagents and samples
    • Ultra-smooth vessel interiors minimize enzyme nucleic acid binding
  • Precision Thermal Performance
    • Thin, uniform walls ensure consistent, precise thermal transfer for reliable results
    • Designed for rapid and accurate heat transfer, reducing cycle times
  • Wide Compatibility
    • Suitable for most commercially available thermal cyclers
    • White PCR plates, enhanced with titanium dioxide (TiO2), optimize fluorescence signal reflection and are recommended for qPCR applications
  • User-Friendly Features
    • Blue alphanumeric coding for easy sample identification
    • Well rims designed to prevent cross-contamination and ensure reliable sealing with tailored mats
    • Skirted plates provide extra rigidity, ideal for automated pipetting systems and transport
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Binding Property Untreated
Format Standard
Array 16 x 24
Autoclavable Autoclavable
Packaging Bagged
Color Clear
Disposable Single-use
Product Type PCR 384-well plate
For Use With (Equipment) Thermal Cyclers
Skirt Style Fully skirted
Material Polypropylene
Volume (Metric) Well 0.03 mL
No. of Wells 384
Sterility Non-sterile
Surface Treatment Non-treated
Total Volume (Metric) Well 0.03 mL
Well Shape Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) PCR
No. per Pack 50
Quantity 50/Pk.
RNase Free Yes