Learn More
3M™ 7000 Series Half- and Full-Facepiece Respirator Accessory, Nose Cup Valve Assembly 7882
Click to view available options
:
7882 Nose cup valve assembly
Description
- Use with either 3M Nose Cup Assembly 7881S or 3M Nose Cup Assembly 7981S
- Small
- 3M Catalog# 7882
Specifications
Specifications
| Product Type | Nosecup Assembly |
| For Use With (Equipment) | 7000 series half and full facepiece respirator |
| Model | 7882 |
| Dimensions (L x W x H) | 11.2 x 8.6 x 4.8 in. (28.4 x 21.8 x 12.1 cm) |
| Length (English) | 11.2 in. |
| Length (Metric) | 28.4 cm |
| Width (English) | 8.6 in. |
| Width (Metric) | 21.8 cm |
| Height (English) | 4.8 in. |
| Height (Metric) | 12.1 cm |
| Show More |