Learn More
3M KCl Electrolyte Solution, Hamilton™
Product Code. p-8263036
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
500mL
Each
Product Code. 10504555
Brand: Hamilton 238036
Replacement and storage electrolytes for optimal sensor health. Hamilton™ 3M KCl Electrolyte Solution helps ease pH and Redox electrode calibration.
  • Helps with buffer values changing over time
Specifications
Concentration 3 M
For Use With (Equipment) KCL Electrode
Physical Form Liquid
Quantity 100 mL
Solution Type Electrolyte